Product Details
- SNP ID
-
rs189422284
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:99622557 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTGTCCCTGCTCAGCACTACCT[C/G]TTCTGCAGGCGGGTCCTGGCCTCCC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZSCAN25
PubMed Links
Gene Details
- Gene
- ZSCAN25
- Gene Name
- zinc finger and SCAN domain containing 25
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145115.2 |
1180 |
Missense Mutation |
CTT,GTT |
L200V |
NP_660090.2 |
XM_005250194.2 |
1180 |
Missense Mutation |
CTT,GTT |
L200V |
XP_005250251.1 |
XM_011515905.2 |
1180 |
Missense Mutation |
CTT,GTT |
L200V |
XP_011514207.1 |
XM_011515906.2 |
1180 |
Missense Mutation |
CTT,GTT |
L200V |
XP_011514208.1 |
XM_011515907.2 |
1180 |
Missense Mutation |
CTT,GTT |
L200V |
XP_011514209.1 |
XM_011515908.2 |
1180 |
Missense Mutation |
CTT,GTT |
L36V |
XP_011514210.1 |
XM_011515909.2 |
1180 |
Missense Mutation |
CTT,GTT |
L200V |
XP_011514211.1 |
XM_011515910.2 |
1180 |
Missense Mutation |
CTT,GTT |
L200V |
XP_011514212.1 |
XM_017011824.1 |
1180 |
Missense Mutation |
CTT,GTT |
L200V |
XP_016867313.1 |
XM_017011825.1 |
1180 |
Missense Mutation |
CTT,GTT |
L36V |
XP_016867314.1 |
XM_017011826.1 |
1180 |
Missense Mutation |
CTT,GTT |
L36V |
XP_016867315.1 |
XM_017011827.1 |
1180 |
Missense Mutation |
CTT,GTT |
L200V |
XP_016867316.1 |
View Full Product Details