Product Details

SNP ID
rs189174307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102450986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCCGGCTGCGGAAGCTGTGGGC[C/T]TGGACTGGGGGTAGCAGCAGAGGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf95 PubMed Links

Gene Details

Gene
C10orf95
Gene Name
chromosome 10 open reading frame 95
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024886.2 320 Missense Mutation AAG,AGG K82R NP_079162.1
Gene
MFSD13A
Gene Name
major facilitator superfamily domain containing 13A
There are no transcripts associated with this gene.

Gene
RPARP-AS1
Gene Name
RPARP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details