Product Details

SNP ID
rs188944294
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:1072350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCCAGGCACTCCAGGCAGAAGA[C/T]GTGGGTGCAGGAGAGCTCCTTGGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf159 PubMed Links

Gene Details

Gene
C1orf159
Gene Name
chromosome 1 open reading frame 159
There are no transcripts associated with this gene.

Gene
LOC100288175
Gene Name
uncharacterized LOC100288175
There are no transcripts associated with this gene.

Gene
LOC105378948
Gene Name
uncharacterized LOC105378948
There are no transcripts associated with this gene.

Gene
RNF223
Gene Name
ring finger protein 223
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205252.1 518 Missense Mutation ATC,GTC I73V NP_001192181.1
XM_017001286.1 518 Missense Mutation ATC,GTC I73V XP_016856775.1
XM_017001287.1 518 Intron XP_016856776.1

View Full Product Details