Product Details

SNP ID
rs188916293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113006725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTAAAGTCGGGGACCAGCCCAC[A/C]CTGCAGCTGAAGGTGAGTGTGGAAG
Phenotype
MIM: 605509
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IL36A PubMed Links

Gene Details

Gene
IL36A
Gene Name
interleukin 36, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014440.2 663 Silent Mutation ACA,ACC T84T NP_055255.1
XM_005263639.2 663 Silent Mutation ACA,ACC T84T XP_005263696.1
XM_011510965.1 663 Silent Mutation ACA,ACC T84T XP_011509267.1
XM_017003806.1 663 Silent Mutation ACA,ACC T84T XP_016859295.1

View Full Product Details