Product Details

SNP ID
rs189560102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:91409384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCAGAGATTGGTTGTGTGGCTA[A/T]AAGTCTCTTGAGTGAGACAACCTCT
Phenotype
MIM: 605327
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NFIL3 PubMed Links

Gene Details

Gene
NFIL3
Gene Name
nuclear factor, interleukin 3 regulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289999.1 1884 Missense Mutation ATA,TTA I451L NP_001276928.1
NM_001290000.1 1884 Missense Mutation ATA,TTA I451L NP_001276929.1
NM_005384.2 1884 Missense Mutation ATA,TTA I451L NP_005375.2
XM_017014743.1 1884 Missense Mutation ATA,TTA I451L XP_016870232.1
XM_017014744.1 1884 Missense Mutation ATA,TTA I451L XP_016870233.1

View Full Product Details