Product Details

SNP ID
rs190259697
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84222111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCACTTGCTGCAGCTGCAGGGGC[C/T]CTGGGCTGCCCAGGCATAGGCGCCC
Phenotype
MIM: 609502 MIM: 616103
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CDHR1 PubMed Links
Additional Information
For this assay, SNP(s) [rs12218853,rs74145770] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CDHR1
Gene Name
cadherin related family member 1
There are no transcripts associated with this gene.

Gene
LRIT1
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 1
There are no transcripts associated with this gene.

Gene
LRIT2
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017924.3 1531 Missense Mutation AGC,GGC S488G NP_001017924.1
NM_001284223.1 1531 Missense Mutation AGC,GGC S498G NP_001271152.1

View Full Product Details