Product Details

SNP ID
rs191028771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9757614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAACGTTAAGGTATGTGGAATAC[C/T]TGAAGGCTTTCCCACATTCTTTACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF846 PubMed Links

Gene Details

Gene
ZNF846
Gene Name
zinc finger protein 846
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077624.1 2434 Missense Mutation NP_001071092.1
XM_005259772.3 2434 Missense Mutation XP_005259829.1
XM_005259773.4 2434 Missense Mutation XP_005259830.1
XM_005259775.3 2434 Missense Mutation XP_005259832.1
XM_006722658.3 2434 Intron XP_006722721.1
XM_011527717.2 2434 Missense Mutation XP_011526019.1
XM_011527718.2 2434 Missense Mutation XP_011526020.1
XM_011527719.2 2434 Missense Mutation XP_011526021.1
XM_011527720.2 2434 Missense Mutation XP_011526022.1
XM_011527721.2 2434 Missense Mutation XP_011526023.1
XM_017026404.1 2434 Missense Mutation XP_016881893.1
XM_017026405.1 2434 Missense Mutation XP_016881894.1
XM_017026406.1 2434 Missense Mutation XP_016881895.1
XM_017026407.1 2434 Intron XP_016881896.1
XM_017026408.1 2434 Intron XP_016881897.1

View Full Product Details