Product Details

SNP ID
rs190990021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102653965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCAGGTCAAGCCTGAAAAGAAG[C/G]CTGGGGAAGAGGTTGTAGGTGGGGC
Phenotype
MIM: 300417
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARMCX5-GPRASP2 PubMed Links

Gene Details

Gene
ARMCX5-GPRASP2
Gene Name
ARMCX5-GPRASP2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199818.1 777 Intron NP_001186747.1
Gene
GPRASP1
Gene Name
G protein-coupled receptor associated sorting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099410.1 777 Missense Mutation CCT,GCT P18A NP_001092880.1
NM_001099411.1 777 Missense Mutation CCT,GCT P18A NP_001092881.1
NM_001184727.1 777 Missense Mutation CCT,GCT P18A NP_001171656.1
NM_014710.4 777 Missense Mutation CCT,GCT P18A NP_055525.3
XM_017029981.1 777 Missense Mutation CCT,GCT P18A XP_016885470.1
XM_017029982.1 777 Missense Mutation CCT,GCT P18A XP_016885471.1

View Full Product Details