Product Details

SNP ID
rs190867203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102729874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAGATCTGGCCTTCTTTCTTTC[C/T]TGGCTTTCTGGGTGCCTGTGAGACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC168 PubMed Links

Gene Details

Gene
CCDC168
Gene Name
coiled-coil domain containing 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146197.1 20963 Silent Mutation CAA,CAG Q6941Q NP_001139669.1
XM_011521106.1 20963 Silent Mutation CAA,CAG Q6901Q XP_011519408.1

View Full Product Details