Product Details
- SNP ID
-
rs191243474
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:128942255 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACAAGGCATGGTCCCAGCCAGGA[C/T]GGAGATAACACCATCTTCAAGGTAA
- Phenotype
-
MIM: 607218
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IRF5
PubMed Links
Gene Details
- Gene
- IRF5
- Gene Name
- interferon regulatory factor 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098627.3 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
NP_001092097.2 |
NM_001098629.2 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
NP_001092099.1 |
NM_001098630.2 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
NP_001092100.1 |
NM_001242452.2 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
NP_001229381.1 |
NM_032643.4 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
NP_116032.1 |
XM_005250317.3 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
XP_005250374.1 |
XM_006715974.2 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
XP_006716037.1 |
XM_011516158.2 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
XP_011514460.1 |
XM_011516159.2 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
XP_011514461.1 |
XM_011516160.1 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
XP_011514462.1 |
XM_011516161.1 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
XP_011514463.1 |
XM_011516162.1 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
XP_011514464.1 |
XM_011516163.2 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
XP_011514465.1 |
XM_011516164.1 |
412 |
Silent Mutation |
GAC,GAT |
D58D |
XP_011514466.1 |
View Full Product Details