Product Details
- SNP ID
-
rs192069715
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41584995 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCACTTGGTGCGGAAGTCATCCGC[A/G]GCCAGACGGGCATTGTCAATCTGCA
- Phenotype
-
MIM: 148066
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs71373429] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT14
- Gene Name
- keratin 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000526.4 |
|
Intron |
|
|
NP_000517.2 |
View Full Product Details