Product Details

SNP ID
rs191651118
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:244411354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCATTAACAGGTAGTTCTTTAAA[C/T]GCCCTTGCATTTGATATGTCTGTGT
Phenotype
MIM: 103060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADSS PubMed Links

Gene Details

Gene
ADSS
Gene Name
adenylosuccinate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126.3 2240 Silent Mutation GCA,GCG A417A NP_001117.2
XM_011544108.2 2240 Silent Mutation GCA,GCG A357A XP_011542410.1

View Full Product Details