Product Details

SNP ID
rs447972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41159708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAGATAAATATTTGTTTAAAAT[G/T]GAATAGCTCCATATGAGAAATTATG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP4-3 PubMed Links

Gene Details

Gene
KRTAP4-3
Gene Name
keratin associated protein 4-3
There are no transcripts associated with this gene.

Gene
KRTAP4-4
Gene Name
keratin associated protein 4-4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032524.1 1024 UTR 3 NP_115913.1
Gene
KRTAP4-5
Gene Name
keratin associated protein 4-5
There are no transcripts associated with this gene.

View Full Product Details