Product Details

SNP ID
rs5742966
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189788724 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAAATCATTTCATCCTATAATCA[A/G]TGTAAAAAATTGATATGATTTTTAT
Phenotype
MIM: 610073 MIM: 600258
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ORMDL1 PubMed Links
Additional Information
For this assay, SNP(s) [rs5742965] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ORMDL1
Gene Name
ORMDL sphingolipid biosynthesis regulator 1
There are no transcripts associated with this gene.

Gene
PMS1
Gene Name
PMS1 homolog 1, mismatch repair system component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000534.4 Intron NP_000525.1
NM_001128143.1 Intron NP_001121615.1
NM_001128144.1 Intron NP_001121616.1
NM_001289408.1 Intron NP_001276337.1
NM_001289409.1 Intron NP_001276338.1
NM_001321044.1 Intron NP_001307973.1
NM_001321045.1 Intron NP_001307974.1
NM_001321046.1 Intron NP_001307975.1
NM_001321047.1 Intron NP_001307976.1
NM_001321048.1 Intron NP_001307977.1
NM_001321049.1 Intron NP_001307978.1
NM_001321051.1 Intron NP_001307980.1
XM_006712596.1 Intron XP_006712659.1
XM_011511356.2 Intron XP_011509658.1
XM_017004344.1 Intron XP_016859833.1
XM_017004345.1 Intron XP_016859834.1
XM_017004346.1 Intron XP_016859835.1
XM_017004347.1 Intron XP_016859836.1
XM_017004348.1 Intron XP_016859837.1
XM_017004349.1 Intron XP_016859838.1
XM_017004350.1 Intron XP_016859839.1
XM_017004351.1 Intron XP_016859840.1

View Full Product Details