Product Details

SNP ID
rs5991705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:51806760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAATACTGCCAGCCAGCTTTCCA[A/G]GGTGATTGTACGAGAAGACTCCTAA
Phenotype
MIM: 300224
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGED1 PubMed Links

Gene Details

Gene
MAGED1
Gene Name
MAGE family member D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005332.1 Intron NP_001005332.1
NM_001005333.1 Intron NP_001005333.1
NM_006986.3 Intron NP_008917.3
XM_011530835.2 Intron XP_011529137.1
XM_017029978.1 Intron XP_016885467.1
XM_017029979.1 Intron XP_016885468.1
XM_017029980.1 Intron XP_016885469.1

View Full Product Details