Product Details

SNP ID
rs6686944
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:45690636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATGACCTTCCAGTTTTGTTTTG[G/T]TTTTTTTTTTAATTTCACAAAAACA
Phenotype
MIM: 147485
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPBP1L1 PubMed Links

Gene Details

Gene
GPBP1L1
Gene Name
GC-rich promoter binding protein 1 like 1
There are no transcripts associated with this gene.

Gene
IPP
Gene Name
intracisternal A particle-promoted polypeptide
There are no transcripts associated with this gene.

Gene
TMEM69
Gene Name
transmembrane protein 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016486.3 Intron NP_057570.2

View Full Product Details