Product Details

SNP ID
rs7306393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:389875 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCACTCTGCCACTTTATACCCGC[C/T]TCTTCTCCATCAAAACAAGTGCAGG
Phenotype
MIM: 180202
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC77 PubMed Links

Gene Details

Gene
CCDC77
Gene Name
coiled-coil domain containing 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130146.1 Intron NP_001123618.1
NM_001130147.1 Intron NP_001123619.1
NM_001130148.1 Intron NP_001123620.1
NM_032358.3 Intron NP_115734.1
Gene
KDM5A
Gene Name
lysine demethylase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042603.2 Intron NP_001036068.1

View Full Product Details