Product Details

SNP ID
rs7970258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:13084807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTATTCGTAGCCTCTAAAAACC[A/G]TGCTCAAGAGACGGATGTTGGCTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM234B PubMed Links

Gene Details

Gene
FAM234B
Gene Name
family with sequence similarity 234 member B
There are no transcripts associated with this gene.

Gene
GSG1
Gene Name
germ cell associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080554.2 1121 UTR 3 NP_001074023.1
NM_001080555.2 1121 UTR 3 NP_001074024.1
NM_001206842.1 1121 UTR 3 NP_001193771.1
NM_001206843.1 1121 UTR 3 NP_001193772.1
NM_001206845.1 1121 UTR 3 NP_001193774.1
NM_031289.3 1121 UTR 3 NP_112579.2
NM_153823.3 1121 UTR 3 NP_722545.2
XM_005253493.2 1121 Intron XP_005253550.1
XM_005253495.1 1121 Intron XP_005253552.1
XM_011520858.1 1121 Intron XP_011519160.1
XM_011520859.1 1121 Intron XP_011519161.1
XM_011520860.1 1121 Intron XP_011519162.1
XM_011520861.1 1121 Intron XP_011519163.1
XM_011520862.1 1121 Intron XP_011519164.1
XM_011520863.1 1121 Intron XP_011519165.1

View Full Product Details