Product Details

SNP ID
hCV189536683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61818427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAACAACTTGGCAGTGTCTAATG[A/C]GAGCTACAAAACATTTTCTATTCTT
Phenotype
MIM: 606148 MIM: 606149
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FADS1 PubMed Links

Gene Details

Gene
FADS1
Gene Name
fatty acid desaturase 1
There are no transcripts associated with this gene.

Gene
FADS2
Gene Name
fatty acid desaturase 2
There are no transcripts associated with this gene.

Gene
MIR1908
Gene Name
microRNA 1908
There are no transcripts associated with this gene.

View Full Product Details