Product Details

SNP ID
rs7926793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61818084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTGGTATTTGGGCACTTCAAAG[A/G]GAAAGGAAACATAACTGTCCCAAGC
Phenotype
MIM: 606148 MIM: 606149
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FADS1 PubMed Links

Gene Details

Gene
FADS1
Gene Name
fatty acid desaturase 1
There are no transcripts associated with this gene.

Gene
FADS2
Gene Name
fatty acid desaturase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281501.1 Intron NP_001268430.1
NM_001281502.1 Intron NP_001268431.1
NM_004265.3 Intron NP_004256.1
Gene
MIR1908
Gene Name
microRNA 1908
There are no transcripts associated with this gene.

View Full Product Details