Product Details

SNP ID
rs200339261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:21515574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAGGCCCTGACTTTGAGCCAGT[C/G]GATTTAAGGAAGGAGTTTCTTTTTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SKIDA1 PubMed Links

Gene Details

Gene
SKIDA1
Gene Name
SKI/DACH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207371.3 3608 Missense Mutation CCA,CGA P750R NP_997254.3
XM_005252450.3 3608 Missense Mutation CCA,CGA P750R XP_005252507.1
XM_011519480.2 3608 Missense Mutation CCA,CGA P750R XP_011517782.1
XM_017016225.1 3608 Missense Mutation CCA,CGA P750R XP_016871714.1
XM_017016226.1 3608 Missense Mutation CCA,CGA P750R XP_016871715.1

View Full Product Details