Product Details

SNP ID
rs200933834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93500127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTAGTAACTCCAAATCCGAAACT[A/G]CATTAGAAAAATTAAAGGGAGAAAT
Phenotype
MIM: 610000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP55 PubMed Links

Gene Details

Gene
CEP55
Gene Name
centrosomal protein 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127182.1 383 Missense Mutation ACA,GCA T26A NP_001120654.1
NM_018131.4 383 Missense Mutation ACA,GCA T26A NP_060601.3
XM_011539918.1 383 UTR 5 XP_011538220.1
XM_011539919.1 383 Intron XP_011538221.1
XM_011539920.2 383 Intron XP_011538222.1
XM_017016372.1 383 Intron XP_016871861.1
XM_017016373.1 383 Intron XP_016871862.1

View Full Product Details