Product Details

SNP ID
rs201692365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123149661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACTCGGACTTTAAAGAAGAGAA[A/C]GAGAGGCTCCTGCCCGCGGGCTCGC
Phenotype
MIM: 603719 MIM: 600647
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BUB3 PubMed Links

Gene Details

Gene
BUB3
Gene Name
BUB3, mitotic checkpoint protein
There are no transcripts associated with this gene.

Gene
HMX2
Gene Name
H6 family homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005519.1 617 Silent Mutation AAA,AAC K120N NP_005510.1
XM_005269743.3 617 Missense Mutation AAA,AAC K120N XP_005269800.1

View Full Product Details