Product Details

SNP ID
rs200748019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1584508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGGCAACAGGATGACCCACAGC[A/C]TGAGGAACAGCAGCAGGGCTTACAG
Phenotype
MIM: 148022
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP5-1 PubMed Links

Gene Details

Gene
KRTAP5-1
Gene Name
keratin associated protein 5-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005922.1 776 Missense Mutation NP_001005922.1
Gene
KRTAP5-AS1
Gene Name
KRTAP5-1/KRTAP5-2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details