Product Details

SNP ID
rs200714842
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62666419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTCACTGGCTGATAGTTGCCTG[A/G]CGGACCGCTGTCTCTGGGATCGGCT
Phenotype
MIM: 616097
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C11orf98 PubMed Links
Additional Information
For this assay, SNP(s) [rs531793220] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C11orf98
Gene Name
chromosome 11 open reading frame 98
There are no transcripts associated with this gene.

Gene
LBHD1
Gene Name
LBH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024099.3 719 Intron NP_077004.2
Gene
METTL12
Gene Name
methyltransferase like 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001043229.1 719 Missense Mutation ACG,GCG T31A NP_001036694.1
XM_005274232.4 719 Missense Mutation ACG,GCG T154A XP_005274289.1
Gene
SNORA57
Gene Name
small nucleolar RNA, H/ACA box 57
There are no transcripts associated with this gene.

Gene
UQCC3
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 3
There are no transcripts associated with this gene.

View Full Product Details