Product Details

SNP ID
rs200412632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125676389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATCTTCTTTAACATGCACTGCT[A/G]GGAATTCTGAGTGATGCAGGTTCCC
Phenotype
MIM: 102525 MIM: 603078
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACRV1 PubMed Links

Gene Details

Gene
ACRV1
Gene Name
acrosomal vesicle protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001612.5 704 Nonsense Mutation CAG,TAG Q215* NP_001603.1
NM_020069.4 704 Nonsense Mutation CAG,TAG Q196* NP_064454.1
NM_020107.4 704 Nonsense Mutation CAG,TAG Q160* NP_064492.1
NM_020108.4 704 Nonsense Mutation CAG,TAG Q145* NP_064493.1
Gene
CHEK1
Gene Name
checkpoint kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114121.2 704 Intron NP_001107593.1
NM_001114122.2 704 Intron NP_001107594.1
NM_001244846.1 704 Intron NP_001231775.1
NM_001274.5 704 Intron NP_001265.2
XM_011542560.2 704 Intron XP_011540862.1
XM_011542561.2 704 Intron XP_011540863.1
XM_011542562.2 704 Intron XP_011540864.1
XM_011542563.2 704 Intron XP_011540865.1
XM_017017146.1 704 Intron XP_016872635.1

View Full Product Details