Product Details

SNP ID
rs199955107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13492587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGCACATCCTGCAGCTTCTTAC[A/G]CAGCCATTCTACTCTCTCCATCGAG
Phenotype
MIM: 168450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTH PubMed Links

Gene Details

Gene
PTH
Gene Name
parathyroid hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000315.3 305 Missense Mutation CGT,TGT R56C NP_000306.1
NM_001316352.1 305 Missense Mutation CGT,TGT R88C NP_001303281.1

View Full Product Details