Product Details
- SNP ID
-
rs199639598
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:62807559 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCACCGGTTGGAGGTGACAGACTG[C/G]AAGTACTTGAGGATCTCTGAATGGG
- Phenotype
-
MIM: 602647
MIM: 603189
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NXF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2228626] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NXF1
- Gene Name
- nuclear RNA export factor 1
There are no transcripts associated with this gene.
- Gene
- STX5
- Gene Name
- syntaxin 5
View Full Product Details