Product Details

SNP ID
rs199666978
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.11:55572207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTTGGAAGAAAATAGTGTTTG[C/T]TATTTTTTTGCGTCTCTACTTGGGA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR4C16 PubMed Links

Gene Details

Gene
OR4C16
Gene Name
olfactory receptor family 4 subfamily C member 16 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004701.2 80 Missense Mutation GCT,GTT A27V NP_001004701.2

View Full Product Details