Product Details

SNP ID
rs201754988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64224797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTCTCGCACTTGTCCCCTCACCC[C/T]GACCTGCAGGGAATGGCCCTGGTTG
Phenotype
MIM: 604189 MIM: 607901 MIM: 610470 MIM: 601398
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC4 PubMed Links

Gene Details

Gene
DNAJC4
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
There are no transcripts associated with this gene.

Gene
FERMT3
Gene Name
fermitin family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031471.5 545 Intron NP_113659.3
NM_178443.2 545 Intron NP_848537.1
XM_011545294.2 545 Intron XP_011543596.1
XM_011545295.2 545 Intron XP_011543597.1
XM_017018398.1 545 Intron XP_016873887.1
XM_017018399.1 545 Intron XP_016873888.1
Gene
NUDT22
Gene Name
nudix hydrolase 22
There are no transcripts associated with this gene.

Gene
TRPT1
Gene Name
tRNA phosphotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033678.3 545 Intron NP_001028850.2
NM_001160389.1 545 Missense Mutation NP_001153861.1
NM_001160390.1 545 Intron NP_001153862.1
NM_001160392.1 545 Intron NP_001153864.1
NM_001160393.1 545 Intron NP_001153865.1
NM_031472.3 545 Intron NP_113660.1
XM_005274345.4 545 Intron XP_005274402.1
XM_005274346.4 545 Intron XP_005274403.1
XM_005274347.4 545 Intron XP_005274404.1
XM_006718706.3 545 Intron XP_006718769.1
XM_006718707.3 545 Intron XP_006718770.1
XM_017018400.1 545 Intron XP_016873889.1
XM_017018401.1 545 Intron XP_016873890.1
Gene
VEGFB
Gene Name
vascular endothelial growth factor B
There are no transcripts associated with this gene.

View Full Product Details