Product Details

SNP ID
rs201603322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65376538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCTGCCGGATGCTGCTCACCA[G/T]GCAGTCCAGCATCCCCTGGTACACT
Phenotype
MIM: 610825
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A45 PubMed Links

Gene Details

Gene
SLC25A45
Gene Name
solute carrier family 25 member 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077241.2 1745 Missense Mutation NP_001070709.2
NM_001278250.2 1745 Missense Mutation NP_001265179.2
NM_001278251.2 1745 Missense Mutation NP_001265180.2
NM_001300820.1 1745 Missense Mutation NP_001287749.1
NM_182556.3 1745 Missense Mutation NP_872362.3
XM_006718509.3 1745 Missense Mutation XP_006718572.1
XM_006718510.3 1745 Missense Mutation XP_006718573.1
XM_011544943.2 1745 Missense Mutation XP_011543245.1
XM_011544944.2 1745 Missense Mutation XP_011543246.1
XM_011544947.2 1745 Missense Mutation XP_011543249.1
XM_011544949.2 1745 Missense Mutation XP_011543251.1
XM_017017562.1 1745 Missense Mutation XP_016873051.1
XM_017017563.1 1745 Missense Mutation XP_016873052.1
XM_017017564.1 1745 Missense Mutation XP_016873053.1
XM_017017565.1 1745 Missense Mutation XP_016873054.1
XM_017017566.1 1745 Missense Mutation XP_016873055.1
XM_017017567.1 1745 Missense Mutation XP_016873056.1
XM_017017568.1 1745 Missense Mutation XP_016873057.1
XM_017017569.1 1745 Missense Mutation XP_016873058.1
XM_017017570.1 1745 Missense Mutation XP_016873059.1
XM_017017571.1 1745 Missense Mutation XP_016873060.1

View Full Product Details