Product Details

SNP ID
rs201605703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77039907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCCGGCGGCCGAGGCGTGTTCC[C/T]GCTCCTGGCGGTGAAGTCGGCGCCT
Phenotype
MIM: 615315
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GNT6 PubMed Links

Gene Details

Gene
B3GNT6
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138706.4 494 Missense Mutation CCG,CTG P119L NP_619651.3

View Full Product Details