Product Details

SNP ID
rs202036216
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74457349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTATCCCAGGATGAGGCTGCCC[A/C]CAGTTACAGCAAATAGAAACATGAC
Phenotype
MIM: 604433
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
KCNE3 PubMed Links

Gene Details

Gene
KCNE3
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005472.4 632 Missense Mutation GGG,GTG G72V NP_005463.1
XM_011544713.2 632 Missense Mutation GGG,GTG G116V XP_011543015.1
XM_017017047.1 632 Missense Mutation GGG,GTG G72V XP_016872536.1
XM_017017048.1 632 Missense Mutation GGG,GTG G72V XP_016872537.1
XM_017017049.1 632 Missense Mutation GGG,GTG G72V XP_016872538.1
XM_017017050.1 632 Missense Mutation GGG,GTG G72V XP_016872539.1
XM_017017051.1 632 Missense Mutation GGG,GTG G72V XP_016872540.1
XM_017017052.1 632 Missense Mutation GGG,GTG G72V XP_016872541.1

View Full Product Details