Product Details

SNP ID
rs901493547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:96006319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTTGAGCTAGTACTTACCCTCT[G/A]GAGCGTCTGTGCTAAAAACTCATTC
Phenotype
MIM: 609457 MIM: 151570
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
HAL PubMed Links

Gene Details

Gene
HAL
Gene Name
histidine ammonia-lyase
There are no transcripts associated with this gene.

Gene
LTA4H
Gene Name
leukotriene A4 hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000895.2 1598 Nonsense Mutation CAG,TAG Q509* NP_000886.1
NM_001256643.1 1598 Nonsense Mutation CAG,TAG Q485* NP_001243572.1
NM_001256644.1 1598 Nonsense Mutation CAG,TAG Q485* NP_001243573.1

View Full Product Details