Product Details

SNP ID
rs200186819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:25215521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATTTTTTTCAATCTGTATTGTC[A/G]GATCTCTCTCACCAATGTATAAAAA
Phenotype
MIM: 190070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRAS PubMed Links

Gene Details

Gene
KRAS
Gene Name
KRAS proto-oncogene, GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004985.4 688 Intron NP_004976.2
NM_033360.3 688 Nonsense Mutation CGA,TGA R164* NP_203524.1
XM_006719069.3 688 Nonsense Mutation CGA,TGA R164* XP_006719132.1
XM_011520653.2 688 Intron XP_011518955.1
XM_017019292.1 688 Intron XP_016874781.1
XM_017019293.1 688 Nonsense Mutation CGA,TGA R164* XP_016874782.1

View Full Product Details