Product Details
- SNP ID
-
rs201891084
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:13088870 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTAGTCCTTTCTCACCTCTCTTG[A/G]CTGGTGGTGTAAGTTCAATGAAACT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM234B
PubMed Links
Gene Details
- Gene
- FAM234B
- Gene Name
- family with sequence similarity 234 member B
There are no transcripts associated with this gene.
- Gene
- GSG1
- Gene Name
- germ cell associated 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080554.2 |
711 |
Missense Mutation |
GCC,GTC |
A158V |
NP_001074023.1 |
NM_001080555.2 |
711 |
Missense Mutation |
GCC,GTC |
A158V |
NP_001074024.1 |
NM_001206842.1 |
711 |
Missense Mutation |
GCC,GTC |
A135V |
NP_001193771.1 |
NM_001206843.1 |
711 |
Missense Mutation |
GCC,GTC |
A145V |
NP_001193772.1 |
NM_001206845.1 |
711 |
Missense Mutation |
GCC,GTC |
A122V |
NP_001193774.1 |
NM_031289.3 |
711 |
Missense Mutation |
GCC,GTC |
A122V |
NP_112579.2 |
NM_153823.3 |
711 |
Missense Mutation |
GCC,GTC |
A122V |
NP_722545.2 |
XM_005253493.2 |
711 |
Missense Mutation |
GCC,GTC |
A182V |
XP_005253550.1 |
XM_005253495.1 |
711 |
Missense Mutation |
GCC,GTC |
A182V |
XP_005253552.1 |
XM_011520858.1 |
711 |
Missense Mutation |
GCC,GTC |
A182V |
XP_011519160.1 |
XM_011520859.1 |
711 |
Missense Mutation |
GCC,GTC |
A158V |
XP_011519161.1 |
XM_011520860.1 |
711 |
Missense Mutation |
GCC,GTC |
A182V |
XP_011519162.1 |
XM_011520861.1 |
711 |
Missense Mutation |
GCC,GTC |
A135V |
XP_011519163.1 |
XM_011520862.1 |
711 |
Missense Mutation |
GCC,GTC |
A158V |
XP_011519164.1 |
XM_011520863.1 |
711 |
Missense Mutation |
GCC,GTC |
A135V |
XP_011519165.1 |
View Full Product Details