Product Details

SNP ID
rs202245183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:100200852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTGCGGTGTGGTTGGTGGTGAG[A/T]TGACGACCTTAGTGCTGGATAATGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ACTR6 PubMed Links

Gene Details

Gene
ACTR6
Gene Name
ARP6 actin-related protein 6 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022496.4 766 Missense Mutation ATG,TTG M1L NP_071941.1
XM_017019821.1 766 UTR 5 XP_016875310.1
Gene
DEPDC4
Gene Name
DEP domain containing 4
There are no transcripts associated with this gene.

View Full Product Details