Product Details

SNP ID
rs202026087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:100204941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTATTTGTTTCCTTGTTTCTAGG[G/T]TTATTCCTAATTGTCAGTTCCGGTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACTR6 PubMed Links

Gene Details

Gene
ACTR6
Gene Name
ARP6 actin-related protein 6 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022496.4 835 Missense Mutation GTT,TTT V24F NP_071941.1
XM_017019821.1 835 UTR 5 XP_016875310.1
Gene
DEPDC4
Gene Name
DEP domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319310.1 835 Intron NP_001306239.1
NM_001319311.1 835 Intron NP_001306240.1
NM_152317.3 835 Intron NP_689530.1
XM_011537876.2 835 Intron XP_011536178.1
XM_017018779.1 835 Intron XP_016874268.1
XM_017018780.1 835 Intron XP_016874269.1
XM_017018781.1 835 Intron XP_016874270.1
XM_017018782.1 835 Intron XP_016874271.1
XM_017018783.1 835 Intron XP_016874272.1
XM_017018784.1 835 Intron XP_016874273.1
XM_017018785.1 835 Intron XP_016874274.1

View Full Product Details