Product Details

SNP ID
rs201417215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56318331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGAGCACCTTGTGGAAAGACTC[A/G]GGCTCAGTGCCATTTTTGCTTAGCT
Phenotype
MIM: 605861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPY2 PubMed Links

Gene Details

Gene
CNPY2
Gene Name
canopy FGF signaling regulator 2
There are no transcripts associated with this gene.

Gene
PAN2
Gene Name
PAN2 poly(A) specific ribonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127460.2 3857 Silent Mutation CCC,CCT P1160P NP_001120932.1
NM_001166279.1 3857 Silent Mutation CCC,CCT P1159P NP_001159751.1
NM_014871.4 3857 Silent Mutation CCC,CCT P1156P NP_055686.3

View Full Product Details