Product Details

SNP ID
rs201428558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41131330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGGCTGAGCAGCTAGATAGAT[A/G]TCATGGTCAGGAGAGATACAGACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KBTBD6 PubMed Links

Gene Details

Gene
KBTBD6
Gene Name
kelch repeat and BTB domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152903.4 1471 Silent Mutation GAC,GAT D394D NP_690867.3
Gene
LOC101929140
Gene Name
uncharacterized LOC101929140
There are no transcripts associated with this gene.

View Full Product Details