Product Details

SNP ID
rs200666130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49705640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTACCTCCACATTCCTCTATTAT[A/T]TCAGCTATTGTGCTTGCTTCATCAC
Phenotype
MIM: 601892
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KPNA3 PubMed Links

Gene Details

Gene
KPNA3
Gene Name
karyopherin subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002267.3 1451 Missense Mutation GAA,GAT E451D NP_002258.2
XM_017020561.1 1451 Missense Mutation GAA,GAT E427D XP_016876050.1

View Full Product Details