Product Details

SNP ID
rs201578280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24606611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTCTCATGCCTGCTGTTAGAGGC[A/G]CTTCATTGTTCTCTTTATCCAGGGC
Phenotype
MIM: 116831
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GZMH PubMed Links

Gene Details

Gene
GZMH
Gene Name
granzyme H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270780.1 582 Missense Mutation CGC,TGC R181C NP_001257709.1
NM_001270781.1 582 Missense Mutation CGC,TGC R159C NP_001257710.1
NM_033423.4 582 Missense Mutation CGC,TGC R245C NP_219491.1
XM_011536683.2 582 Missense Mutation CGC,TGC R211C XP_011534985.1

View Full Product Details