Product Details

SNP ID
rs199784429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24606657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCAGGAAGTGTGAGACCTTGAT[A/G]TAGACTCCTGGAGGTGTCCCTTTTT
Phenotype
MIM: 116831
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GZMH PubMed Links

Gene Details

Gene
GZMH
Gene Name
granzyme H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270780.1 536 Silent Mutation TAC,TAT Y165Y NP_001257709.1
NM_001270781.1 536 Silent Mutation TAC,TAT Y143Y NP_001257710.1
NM_033423.4 536 Silent Mutation TAC,TAT Y229Y NP_219491.1
XM_011536683.2 536 Silent Mutation TAC,TAT Y195Y XP_011534985.1

View Full Product Details