Product Details

SNP ID
rs199685581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94127978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGAGGATGTTGGATGATGTGG[A/C]GAGTCCAGCTGCCCCTGTGGAGGAG
Phenotype
MIM: 611319
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IFI27L2 PubMed Links

Gene Details

Gene
IFI27L2
Gene Name
interferon alpha inducible protein 27 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032036.2 270 Missense Mutation GCC,TCC A72S NP_114425.1

View Full Product Details