Product Details

SNP ID
rs199882985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:72971052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTGAACTCCTTCCGGCAGTTGT[A/G]GCAGTGGAGGATTTCGTGGTCAGGC
Phenotype
MIM: 616372 MIM: 605471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF4 PubMed Links

Gene Details

Gene
DCAF4
Gene Name
DDB1 and CUL4 associated factor 4
There are no transcripts associated with this gene.

Gene
ZFYVE1
Gene Name
zinc finger FYVE-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281734.1 1021 Missense Mutation CAC,TAC H708Y NP_001268663.1
NM_001281735.1 1021 Missense Mutation CAC,TAC H307Y NP_001268664.1
NM_021260.3 1021 Missense Mutation CAC,TAC H722Y NP_067083.1
NM_178441.2 1021 Missense Mutation CAC,TAC H307Y NP_848535.1
XM_017021373.1 1021 Missense Mutation CAC,TAC H307Y XP_016876862.1

View Full Product Details