Product Details

SNP ID
rs200998769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22603390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCATGGGATTCTTTCCTCCCCAG[A/G]TCTCCAGAATAAGTTCCTGGCCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD4 PubMed Links

Gene Details

Gene
ABHD4
Gene Name
abhydrolase domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022060.2 235 Missense Mutation TAT,TGT Y38C NP_071343.2
XM_005267986.4 235 Missense Mutation TAT,TGT Y14C XP_005268043.1

View Full Product Details