Product Details
- SNP ID
-
rs200680738
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:50182700 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTGGACCGGCGCAGCAATCAAGT[A/G]GCCCGGGCGCTGCACGACCACCTCG
- Phenotype
-
MIM: 609123
MIM: 603247
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP8B4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34490804] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP8B4
- Gene Name
- ATPase phospholipid transporting 8B4 (putative)
- Gene
- SLC27A2
- Gene Name
- solute carrier family 27 member 2
View Full Product Details