Product Details

SNP ID
rs199520953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34982489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGAATTTTGCCACTTCTAGCAG[C/T]AAAACAGCTGTGTTCATTGAGAATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF770 PubMed Links

Gene Details

Gene
ZNF770
Gene Name
zinc finger protein 770
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014106.3 Intron NP_054825.2
XM_011521744.2 Intron XP_011520046.1

View Full Product Details