Product Details

SNP ID
rs199716895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40606425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCCCAAGGAGTCCTCTTCAGGA[A/C]CTCAGAGGTGGGAATGAAAGAGTTC
Phenotype
MIM: 609173
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KNL1 PubMed Links

Gene Details

Gene
KNL1
Gene Name
kinetochore scaffold 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144508.4 269 Missense Mutation GAA,GAC E36D NP_653091.3
NM_170589.4 269 Missense Mutation GAA,GAC E36D NP_733468.3
XM_017022432.1 269 UTR 5 XP_016877921.1

View Full Product Details