Product Details
- SNP ID
-
rs199786602
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:77788785 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCGGCGGCGGCGACGGCTCGCAC[A/C]GCCTCGGGGACGCCCAGGAGATGCG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
VAT1L
PubMed Links
Gene Details
- Gene
- VAT1L
- Gene Name
- vesicle amine transport 1-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020927.2 |
222 |
Missense Mutation |
AGC,CGC |
S35R |
NP_065978.1 |
View Full Product Details